Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nitric acid, Environmental Grade Plus, 68-70%

Click to view available options
Quantity:
500mL
7697-37-2
HNO3
63.012
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]
7697-37-2
HNO3
63.012
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]
Color Colorless to Yellow
Quantity 500mL
UN Number UN2031
Merck Index 14,6577
Density 1.41 g/mL
Formula Weight 63.01
Physical Form Liquid, Double Sub-Boiling Quartz Distillation
Percent Purity 68 to 70%
Grade Environmental Plus
Odor Pungent
Boiling Point 121°C
Chemical Name or Material Nitric acid
Show More Show Less

GHS H Statement
H314-H272
Causes severe skin burns and eye damage.
May intensify fire
oxidizer.

P210-P220-P221-P234-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P390-P501c

H272-H290-H314-H331

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: NITRIC ACID

EINECSNumber : 231-714-2

RTECSNumber : QU5775000

TSCA : Yes

Recommended Storage : Ambient temperatures