Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric acid, extra pure, 60% solution in water, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass Bottle
Plastic bottle
7697-37-2,7732-18-5
HNO3
63.012
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]
7697-37-2,7732-18-5
HNO3
63.012
MFCD00011349
GRYLNZFGIOXLOG-UHFFFAOYSA-N
hydrogen nitrate,aqua fortis,azotic acid,salpetersaeure,rfna,acidum nitricum,nital,acide nitrique,nitrous fumes,nitryl hydroxide
944
CHEBI:48107
nitric acid
[N+](=O)(O)[O-]
Color Colorless to Yellow
Quantity 1L
CAS Min % 59.0,37.0
CAS Max % 63.0,41.0
Linear Formula HNO3
Fieser 01,733; 03,212; 04,356; 05,474
Merck Index 15, 6663
Chloride 10ppm max.
Density 1.3300g/mL
Formula Weight 63.01
Physical Form Liquid
Percent Purity 59.0 to 63.0%
Grade Extra Pure
Iron (Fe) 5ppm max.
Packaging Glass Bottle
Solubility Solubility in water: >100 g/100mL (20°C).
Specific Gravity 1.33
Melting Point -42.0°C
Boiling Point 122.0°C
Chemical Name or Material Nitric acid
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage. May be corrosive to metals. Corrosive to the respiratory tract.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

Danger