Nitric Acid, Fuming (Certified ACS), Fisher Chemical

Packaging Glass Bottle
Quantity 500mL

 View more versions of this product

Catalog No. A202-500

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 

Custom packing/Bulk ordering

You can now order this item in custom quantities.  Learn More

    Chemical Identifiers

    CAS 7697-37-2, 7732-18-5, 10102-44-0,7732-18-5,10102-44-0
    Physical Form Liquid
    Assay Percent Range ≥90.0 %
    Molecular Formula HNO3
    Formula Weight 63.01
    MDL Number MFCD00011349
    Chemical Name or Material Nitric Acid
    Assay ≥90.0%
    Grade Certified ACS Grade

    Specifications

    Melting Point −41°C
    Boiling Point 120.5°C
    Formula Weight 63.01
    Quantity 500mL
    CAS 7697-37-2, 7732-18-5, 10102-44-0,10102-44-0,7732-18-5
    UN Number 2032
    Packaging Glass Bottle
    DOT Information DOT Class 8, : Corrosive
    Physical Form Liquid
    Color Clear Colorless Light Yellow
    pH 1.0
    Density 1.5g/mL
    Vapor Pressure 0.37 - 0.40 kPa
    Molecular Formula HNO3
    Health Hazard 3 Emergency Overview
    Oxidizer: Contact with combustible/organic material may cause fire. May cause pulmonary edema. Hygroscopic. Containers should be vented periodically in order to overcome pressure buildup. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

    NFPA

    Health:4
    Flammability:0
    Instability:0
    Assay Percent Range ≥90.0 %
    Chloride 0.7ppm max.
    Dilution Test Pass Test
    Health Hazard 2 DANGER!
    Identification Pass Test
    Additional Information Vapor Pressure: 0.37 to 0.40kPa
    Arsenic (As) 0.3ppm max.
    Cleanup Symbol Acid
    MDL Number MFCD00011349
    Dissolved Oxides 0.10% max.
    Heavy Metals (as Pb) 5ppm max.
    Ignition Residue 0.002% max.
    Iron (Fe) 2ppm max.
    Sulfate 5ppm max.
    ChemAlert Storage Symbol Yellow
    Chemical Name or Material Nitric Acid
    Assay ≥90.0%
    Grade Certified ACS Grade
       

    DANGER!

    Emergency Overview
    Oxidizer: Contact with combustible/organic material may cause fire. May cause pulmonary edema. Hygroscopic. Containers should be vented periodically in order to overcome pressure buildup. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

    NFPA

    Health:4
    Flammability:0
    Instability:0

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Product Certifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit