Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid, TraceGrade, 67-70%, Spectrum™

Click to view available options
Quantity:
2.5 L
6 x 500 mL
4 x 2.5 L
Packaging:
Poly Bottle
7697-37-2
HNO3
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
O[N+]([O-])=O
7697-37-2
HNO3
63.01
GRYLNZFGIOXLOG-UHFFFAOYSA-N
nitric acid
O[N+]([O-])=O

N1084, 7697-37-2, HNO3

Spectrum™ Nitric Acid, TraceGrade is a strong, highly corrosive mineral acid with extremely low metal content (over 50 with levels less than 1 PPB). This grade is primarily used as an analytical reagent in elemental analysis determinations using ICP and AA techniques, digestion of water samples and as a doping agent for organic semiconductors.

Aluminum (Al) 1 ppb
Quantity 6 x 500 mL
CAS Max % 1
Antimony (Sb) 0.5 ppb
Arsenic (As) 0.5 ppb
Barium (Ba) 0.1 ppb
Beryllium (Be) 0.1 ppb
Boron (B) 1 ppb
Cadmium (Cd) 0.5 ppb
Calcium (Ca) 1 ppb
Cerium (Ce) 0.1 ppb
Cobalt (Co) 0.5 ppb
Chromium (Cr) 1 ppb
Chloride 0.2 ppm
Copper (Cu) 0.5 ppb
Percent Purity 67 to 70%
Gallium (Ga) 0.1 ppb
Iron (Fe) 1 ppb
Lead (Pb) 0.1 ppb
Manganese (Mn) 0.1 ppb
Magnesium (Mg) 1 ppb
Mercury (Hg) 0.1 ppb
Molybdenum (Mo) 0.1 ppb
Nickel (Ni) 0.5 ppb
Packaging Poly Bottle
Potassium (K) 1 ppb
Rhodium (Rh) 0.5 ppb
Rubidium (Rb) 0.1 ppb
Selenium (Se) 1 ppb
Sodium (Na) 1 ppb
Strontium (Sr) 0.1 ppb
Thallium (Tl) 0.1 ppb
Tin (Sn) 0.5 ppb
Titanium (Ti) 0.5 ppb
Zinc (Zn) 0.5 ppb
Show More Show Less