Learn More
BioVision IncSupplier Diversity Partner Nitric Oxide Colorimetric Assay Kit: Nitric Oxide Colorimetric Assay Kit: Accurate & Convenient Colorimetric Assay to Measure Nitric Oxide from Urine, Serum etc. Detection Limit: 1 μM. 200 Assays. DFS Item
Manufacturer:  BioVision IncSupplier Diversity Partner K262200
Nitric oxide (NO) plays an important role in neurotransmission, vascular regulation, immune response and apoptosis. NO is rapidly oxidized to nitrite and nitrate which are used to quantitate NO production. BioVision’s Nitric Oxide Colorimetric Assay Kit provides an accurate, convenient measure of total nitrate/nitrite in a simple two-step process. The first step converts nitrate to nitrite utilizing nitrate reductase. The second step uses Griess Reagents to convert nitrite to a deep purple azo compound. The amount of the azochromophore accurately reflects nitric oxide amount in samples. The detection limit of the assay is approximately 0.1 nmole nitrite/well, or 1 µM. BioVision charges an Ice pack and Insulated Container fee of $25.00 for each order.
Catalog No. 50-111-9659
Due to product restrictions, please to purchase or view availability for this product.