Learn More
Honeywell™ NitriGuard Plus™ With Flocked Lining, Blue
Blue nitrile gloves resist punctures and abrasions, while flock lining gives extra comfort working with oils, petroleum solvents or greases.
$4.65 - $48.50
Specifications
Color Blue
Thickness (English) 15 mil
Thickness (Metric) 0.38 mm
Length (English) 13 in.
Length (Metric) 33 cm
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
11-393-64B
View Documents
Honeywell™
LA111EBFL/8
8
Dozen of 12 for $46.75
Case of 12 DZ for $561.00
Add to cart
11-393-64D
View Documents
Honeywell™
LA111EBFL/10
10
Dozen of 12 for $4.65
Save 75% Reg : $18.59
Case of 12 DZ for $55.74
Save 91% Reg : $618.00
Add to cart
11-393-64E
View Documents
Honeywell™
LA111EBFL/11
11
Dozen of 12 for $48.50
Case of 12 DZ for $582.00
Add to cart
11-393-64A
View Documents
Honeywell™
LA111EBFL/7
7 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
11-393-64C
View Documents
Honeywell™
LA111EBFL/9
9 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • 100% nitrile is resistant to cuts, snags, abrasions and punctures
  • Blue with flocked interior
  • 15 mil thickness
  • Curved-hand shape with straight cuff edge
  • For handling fats, petroleum products, and many chemicals
  • USDA accepted for food processing
Specifications
Specifications
Blue
0.38 mm
33 cm
15 mil
13 in.
Raised pattern