Learn More
Honeywell™ NitriGuard Plus™ With Sanitized Interior, Green
Green nitrile gloves feature a sanitized interior and are mechanical and chemical resistant to a large range of products.
$37.25
Specifications
ANSI Abrasion Rating ANSI 2
Color Green
Thickness (English) 11 mil
Thickness (Metric) 0.27 mm
Length (English) 13 in.
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-025-704
View Documents
Honeywell™
LA102G/7
7
Dozen of 12 for $37.25
Case of 12 DZ for $447.00
Add to cart
19-025-705
View Documents
Honeywell™
LA102G/8
8
Dozen of 12 for $37.25
Case of 12 DZ for $447.00
Add to cart
19-025-706
View Documents
Honeywell™
LA102G/9
9
Dozen of 12 for $37.25
Case of 12 DZ for $447.00
Add to cart
19-025707
View Documents
Honeywell™
LA102G/10
10
Dozen of 12 for $37.25
Case of 12 DZ for $447.00
Add to cart
19-025-708
View Documents
Honeywell™
LA102G/11
11
Dozen of 12 for $37.25
Case of 12 DZ for $447.00
Add to cart
Description
Description
  • 100% nitrile is resistant to cuts, snags, abrasions and punctures
  • Green, sanitized interior
  • 11 mil thickness
  • Curved-hand shape with straight cuff edge
  • For handling fats, petroleum products, and many chemicals
  • USDA accepted for food processing
Specifications
Specifications
ANSI 2
11 mil
13 in.
Raised pattern
Green
0.27 mm
33 cm