Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP Nitrile-Coated Gloves with Aramid Fiber
Protect hands without sacrificing comfort or dexterity
$67.69 - $110.00
Specifications
Color | Black, Yellow |
---|---|
For Use With (Application) | Material handling, glass handling operations, fishing industries, sanitation, general maintenance, Woodworking, waste handling and recycling |
Catalog Number | Mfr. No. | Material | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Size | Price | Quantity & Availability | ||||
19-130-3716D
|
PIP
09K1450/L |
Lycra™ | Large |
Dozen of 12 for $84.00
Case of 12 DZ for $1,008.00 |
|
19-130-3716E
|
PIP
09K1450/XL |
Lycra™ | X-Large |
Dozen of 12 for $84.00
Case of 12 DZ for $1,008.00 |
|
19-130-3716C
|
PIP
09K1450/M |
Lycra™ | Medium |
Dozen of 12 for $84.00
Case of 12 DZ for $1,008.00 |
|
19-103-602
|
PIP
09K1660/M |
Nitrile | Medium |
Dozen of 12 for $110.00
Case of 6 DZ for $660.00 |
|
19-103-601
|
PIP
09K1660/S |
Nitrile | Small |
Dozen of 12 for $110.00
Case of 6 DZ for $660.00 |
|
19-103-603
|
PIP
09K1660/L |
Nitrile | Large |
Dozen of 12 for $67.69
Case of 6 DZ for $660.00 |
|
19-103-605
|
PIP
09K1660/XXL |
Nitrile | 2X-Large |
Dozen of 12 for $110.00
Case of 6 DZ for $660.00 |
|
19-103-604
|
PIP
09K1660/XL |
Nitrile | X-Large |
Dozen of 12 for $67.69
Case of 6 DZ for $660.00 |
|
Description
Used for material handling (including glass handling), fishing, sanitation, general maintenance, woodworking waste handling and recycling. Medium weight. Coated palm and fingertips.
- Offer seamless knit construction
- Designed to be cut resistant
- Dupont Kevlar aramid fibers fiber provides high tensile strength relative to its weightup to five times stronger than steel; will not melt, ignite or conduct electricity
- Porous nitrile foam coating provides superior grip in oily applications
Order Info
Cotton color binding denotes sizes
Specifications
Black, Yellow | |
Material handling, glass handling operations, fishing industries, sanitation, general maintenance, Woodworking, waste handling and recycling |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title