Learn More
Nitrile Examination Gloves
Non-sterile, powder-free, nitrile examination gloves.
 View more versions of this product
Catalog No. 12888550B
Add to cart
Description
Description
  • Ambidextrous with beaded cuff
  • Color: Blue
  • Finger textured
  • ASTM D6319
Specifications
Specifications
Blue
Medium
Yes
Palm: 0.06-0.09 mm; Cuff: 0.05-0.08 mm
0.08 to 0.14 mm
Nitrile
ASTM D6319
Palm: 2.4-3.5 mil; Cuff: 2.0-3.1 mil
3.1 to 5.5 mil