Learn More
PIP™ Nitrile Industrial Grade Powder-Free Blue Glove
Economical puncture resistance with great comfort dexterity and tactile sensitivity
$342.50
Specifications
Disposable Yes
Latex Free Yes
Length (English) 10 in.
Material Nitrile
Thickness (English) 4 mil
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-046-451
View Documents
PIP™
29101/S
Small Case of 1000 for $342.50
Add to cart
19-046-452
View Documents
PIP™
29101/M
Medium Case of 1000 for $342.50
Add to cart
19-046-453
View Documents
PIP™
29101/L
Large Case of 1000 for $342.50
Add to cart
19-046-454
View Documents
PIP™
29101/XL
X-Large Case of 1000 for $342.50
Add to cart
Description
Description
  • 4 mil
  • Industrial grade
  • Powder free
  • Blue
  • Economical puncture resistance
  • Great comfort dexterity and tactile sensitivity
  • Textured grip
  • No latex
  • Ambidextrous
  • Rolled cuffs adds additional strength
  • Complies with 21 CFR
Specifications
Specifications
Yes
10 in.
4 mil
Yes
Nitrile
Blue