Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nitrilotriacetic acid, ACS, 98.0% min

Chelating and sequestering agent, used in the manufacture of synthetic detergents

Manufacturer:  Alfa Aesar™ 3651536

Quantity 500g

 View more versions of this product

Catalog No. AA3651536


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

139-13-9
191.14
1710776
Tris(carboxymethyl)amine; N,N-Bis(carboxymethyl)glycine
ACS Grade
N(CH2CO2H)3
MFCD00004287
14,6579
Nitrilotriacetic acid

Specifications

Odorless
246°C (dec.)
139-13-9
N(CH2CO2H)3
MFCD00004287
14,6579
Nitrilotriacetic acid
ACS Grade
White
500g
Powder
191.14
1710776
Tris(carboxymethyl)amine; N,N-Bis(carboxymethyl)glycine
≤98.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H351-H302-H315-H319-H335
Suspected of causing cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AJ0175000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit