Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrilotriacetic acid, trisodium salt, monohydrate, 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
2.5kg
Packaging:
Plastic bottle
18662-53-8
C6H6NNa3O6·H2O
275.09
HERBOKBJKVUALN-UHFFFAOYSA-K
trisodium nitrilotriacetate monohydrate,nitrilotriacetic acid trisodium salt monohydrate,nta sodium hydrate,nta trisodium salt dihydrate,unii-j0u92u6nu5,ccris 437,nitrilotriacetic acid trisodium salt, monohydrate,nitriloacetic acid trisodium salt monohydrate,glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate,nitrilotriacetic acid, trisodium salt monohydrate
29194
trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate
C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+]
18662-53-8
C6H6NNa3O6·H2O
275.09
HERBOKBJKVUALN-UHFFFAOYSA-K
trisodium nitrilotriacetate monohydrate,nitrilotriacetic acid trisodium salt monohydrate,nta sodium hydrate,nta trisodium salt dihydrate,unii-j0u92u6nu5,ccris 437,nitrilotriacetic acid trisodium salt, monohydrate,nitriloacetic acid trisodium salt monohydrate,glycine, n,n-bis carboxymethyl-, trisodium salt, monohydrate,nitrilotriacetic acid, trisodium salt monohydrate
29194
trisodium;2-[bis(carboxylatomethyl)amino]acetate;hydrate
C(C(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.[Na+].[Na+].[Na+]
Infrared Spectrum Authentic
Packaging Plastic bottle
Color White
Quantity 2.5kg
Melting Point >320.0°C
Assay Percent Range 99+%
Linear Formula N(CH2CO2Na)3·H2O
Solubility Information Solubility in water: 484g/L (25°C)
Formula Weight 275.09
Physical Form Powder, Crystals and/or Chunks
Percent Purity ≥99%
Chemical Name or Material Nitrilotriacetic acid, trisodium salt, monohydrate
Show More Show Less