Learn More
Nitrilotriacetic acid, trisodium salt, monohydrate, 99+%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Information
• Nitrilotriacetic acid, trisodium salt, monohydrate is considered to be a nongenotoxic compound with carcinogenic activity
• This compound inhibits gap-junctional communication between Chinese hamster lung fibroblasts
Applications
• Nitrilotriacetic acid trisodium salt monohydrate is commonly used in laboratory experiments due to its capacity to induce the tumorigenic process
• It has been used in in vivo experiments as an inducer of bladder and kidney tumors in animal models
• It is used as a chelating agent, which forms coordination compounds with metal ions (chelates)
Literature References
Malcolm, A.; Mills, L. Inhibition of gap-junctional intercellular communication between Chinese hamster lung fibroblasts by di(2-ethylhexyl) phthalate (DEHP) and trisodium nitrilotriacetate monohydrate (NTA). Cell Biology and Toxicology. 1989, 5, 145–153.
Specifications
Melting Point | >320.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 99+% |
Packaging | Plastic bottle |
Linear Formula | N(CH2CO2Na)3·H2O |
Quantity | 25 g |
Solubility Information | Solubility in water: 484g/L (25°C) |
Formula Weight | 275.09 |
Percent Purity | 99+% |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Suspected of causing cancer. Harmful if swallowed. Causes serious eye irritation. |
Precautionary Statement | GHS P Statement Use personal protective equipment as required. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present an |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.