Learn More
MCR Safety NitriShield™ with Grippaz™ Technology, Orange Nitrile Disposable Gloves, 6 mil, 9.5 inch
6016O NitriShield™ with Grippaz™ Technology, Orange Nitrile, 6 mil, 9.5 inch. The unique fish-scale pattern of these gloves provides unsurpassed grip and durability. Ergonomically designed for wet grip on the outside and to reduce hand slippage on the inside.
Specifications
Back Material Nitrile
Certifications/Compliance FDA Compliant
Coating Material No Coating
Color Orange
Exterior Finish Fish-scale pattern grip
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-812-745
View Documents
MCR Safety
6016OL
Large Case of 1000 for $199.18
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-746
View Documents
MCR Safety
6016OM
Medium Case of 1000 for $199.18
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-747
View Documents
MCR Safety
6016OS
Small Case of 1000 for $199.18
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-748
View Documents
MCR Safety
6016OXL
X-Large Case of 1000 for $199.18
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-749
View Documents
MCR Safety
6016OXXL
XX-Large Case of 1000 for $199.18
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • 6 mil, Orange Powder Free Nitrile
  • Fish-scale pattern grip
  • Textured inside for better traction
  • 9.5 Inch Industrial/food service grade
  • Component materials comply with all relevant federal regulations for food contact
Specifications
Specifications
Nitrile
No Coating
Fish-scale pattern grip
Ambidextrous
Unlined
1 Case (10 boxes of 100)
6 mil
FDA Compliant
Orange
Rolled
Industrial/food service grade
Nitrile
Non-Sterile