Learn More
MCR Safety™ NitriShield™ Nitrile Disposable Gloves
8 mil Nitrile industrial and food service grade gloves
Specifications
Latex Free Yes
Hand Ambidextrous
Material Nitrile
Back Material Nitrile
Certifications/Compliance FDA Compliant
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-812-754
View Documents
MCR Safety™
6030S
Small
Case of 1000 for $330.67
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-755
View Documents
MCR Safety™
6030M
Medium
Case of 1000 for $330.67
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-756
View Documents
MCR Safety™
6030L
Large
Case of 1000 for $330.67
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-757
View Documents
MCR Safety™
6030XL
X-Large
Case of 1000 for $330.67
N/A
Due to product restrictions, please to purchase or view availability for this product.
19-812-758
View Documents
MCR Safety™
6030XXL
2X-Large
Case of 1000 for $330.67
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Nitrile gloves contain no latex proteins and offer excellent resistance to punctures and tears
  • Nitrile gloves are three times more puncture resistant than rubber and can be used to offer superior resistance to many types of chemicals
  • Textured grip
  • This glove is an excellent alternative for individuals who experience allergic reactions to latex
  • The 6030 is an industrial / food service grade powder-free nitrile glove that measures 8 mil in thickness and is 9.5 inches in length
  • Component materials comply with all federal regulations for food contact
  • Specifications
    Specifications
    Yes
    Nitrile
    FDA Compliant
    Blue
    Rolled
    Textured Grip
    Premium
    9.5 in.
    0.20 mm
    Standard
    Ambidextrous
    Nitrile
    Blue
    Blue
    Yes
    Food, Janitorial, Nursing Home Care, Painting
    50/case
    8 mil
    Wing