BioClean P-Zero™ Sterile Polychloroprene Cleanroom Gloves

Excellent chemical protection, tactility and cytotoxic protection

Manufacturer: NITRITEX CANADA BPZS65

 View more versions of this product

Catalog No. 19-668-019

  Check Availability
Add to Cart

    Specifications

    Thickness 7mil
    Certifications ASTM 6978-05, ISO Class 4, EU GMP Grade A
    For Use With (Application) cleanroom environment
    Glove Cuff Style Beaded
    Hand Hand-specific
    Length (English) 12 in.
    Latex Free Yes
    Length (Metric) 30.5cm
    Material Polychloroprene
    Powdered No
    Size 6.5
    Sterile Yes
    Coating Material Textured
    Color Natural

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit