Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stanbio™ Nitro-tab Ketone Tablets
Measure acetoacetic acid and acetone in urine, serum, plasma, and whole blood
Supplier: Stanbio™ 4000100
This item is not returnable.
View return policy
Description
- For the semi-quantitative determination of ketones in urine, serum, plasma, and whole blood
- CLIA waived for urine testing
- Can be stored at room temperature
- Control materials are available
- Urine results in 30 seconds
- Serum and plasma results in 2 minutes
- Whole blood results in 10 minutes
Specifications
Room Temperature (15°C to 30°C) | |
No | |
Ketone | |
Colorimetric | |
Sodium Nitroprusside, Aminoacetic Acid, Disodium Phosphate, Sodum Borate, Lactose, and Non-Reactive Binding Ingredients | |
100/Bottle | |
30 Seconds (Urine), 2 Minutes (Serum/Plasma), 10 Minutes (Whole Blood) |
Yes | |
Waived (Urine) | |
No | |
Ketone Tablets | |
Bottle | |
None | |
Tablet |
Safety and Handling
ShelfLife : 20 Months
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction