Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrobenzene, 99%, Extra Pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
98-95-3
C6H5NO2
123.11
MFCD00007043
LQNUZADURLCDLV-UHFFFAOYSA-N
nitrobenzol,benzene, nitro,oil of mirbane,mirbane oil,essence of mirbane,nitro-benzene,nitrobenzeen,oil of myrbane,nitrobenzen,mononitrobenzene
7416
CHEBI:27798
nitrobenzene
C1=CC=C(C=C1)[N+](=O)[O-]
98-95-3
C6H5NO2
123.11
MFCD00007043
LQNUZADURLCDLV-UHFFFAOYSA-N
nitrobenzol,benzene, nitro,oil of mirbane,mirbane oil,essence of mirbane,nitro-benzene,nitrobenzeen,oil of myrbane,nitrobenzen,mononitrobenzene
7416
CHEBI:27798
nitrobenzene
C1=CC=C(C=C1)[N+](=O)[O-]
Melting Point 5.0°C to 6.0°C
Flash Point 88°C
Boiling Point 210.0°C to 211.0°C
Density 1.2050g/mL
Quantity 1L
Packaging Glass bottle
Color Yellow
Refractive Index 1.5503 to 1.5523
Specific Gravity 1.205
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H5NO2
Beilstein 05, 233
Fieser 02,295; 07,251; 08,358; 17,16
Merck Index 15, 6674
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,benzene,ether and oils
Formula Weight 123.11
Physical Form Liquid
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material Nitrobenzene
Show More Show Less

GHS H Statement
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Toxic in contact with skin.
Harmful to aquatic life with long lasting effects.
Toxic if sw

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove

GHS Signal Word: Danger

EINECSNumber : 202-716-