Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrobenzene, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
98-95-3
C6H5NO2
123.111
LQNUZADURLCDLV-UHFFFAOYSA-N
nitrobenzol,benzene, nitro,oil of mirbane,mirbane oil,essence of mirbane,nitro-benzene,nitrobenzeen,oil of myrbane,nitrobenzen,mononitrobenzene
7416
CHEBI:27798
nitrobenzene
C1=CC=C(C=C1)[N+](=O)[O-]
98-95-3
C6H5NO2
123.111
LQNUZADURLCDLV-UHFFFAOYSA-N
nitrobenzol,benzene, nitro,oil of mirbane,mirbane oil,essence of mirbane,nitro-benzene,nitrobenzeen,oil of myrbane,nitrobenzen,mononitrobenzene
7416
CHEBI:27798
nitrobenzene
C1=CC=C(C=C1)[N+](=O)[O-]

High quality chemicals for laboratory and specialized industrial use

Specific Gravity 1L = 1.20kg
Flash Point 88.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 500mL
Formula Weight 123.11
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material Nitrobenzene