Alfa Aesar™ Nitrobenzene-d5, 99% (Isotopic)

Manufacturer: Alfa Aesar™ 4233706

Quantity 5g

 View more versions of this product

Catalog No. AA4233706

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 4165-60-0
    Formula Weight 128.14
    MDL Number MFCD00044415
    UN Number UN1662
    Chemical Name or Material Nitrobenzene-d5, Isotopic

    Specifications

    Formula Weight 128.14
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H372-H351-H361-H227
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Suspected of damaging fertility or the unborn child.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 211°C
    Quantity 5g
    MDL Number MFCD00044415
    UN Number UN1662
    Density 1.25g/mL
    Melting Point 6°C
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROBENZENE
    Physical Form Liquid
    Flash Point 88°C (190°F)
    CAS 4165-60-0
    Health Hazard 1 Danger
    Chemical Name or Material Nitrobenzene-d5, Isotopic
    Assay 99%

    GHS H Statement
    H300-H310-H330-H372-H351-H361-H227
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes damage to organs through prolonged or repeated exposure.
    Suspected of causing cancer.
    Suspected of damaging fertility or the unborn child.
    Combustible liquid.

    GHS P Statement
    P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit