Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrofurazone, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
100 g
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
59-87-0
C6H6N4O4
198.14
IAIWVQXQOWNYOU-FPYGCLRLSA-N
[(E)-[(5-nitrofuran-2-yl)methylidene]amino]urea
NC(=O)N\N=C\C1=CC=C(O1)[N+]([O-])=O
59-87-0
C6H6N4O4
198.14
IAIWVQXQOWNYOU-FPYGCLRLSA-N
[(E)-[(5-nitrofuran-2-yl)methylidene]amino]urea
NC(=O)N\N=C\C1=CC=C(O1)[N+]([O-])=O

NI140, 59-87-0, C6H6N4O4

Spectrum™ Nitrofurazone, USP is an antibiotic used to treat burns that have become infected. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.001
Loss on Drying 0.005
Quantity 100 g
pH 5.0 to 7.5
CAS Max % 1
Percent Purity 98 to 102%
Grade USP