Learn More
Raylabcon Low Nitrogen Weighing Papers
SDPSCI_ED
Sturdy and nonabsorbent.
Manufacturer:  Raylabcon 20605625/EMD
 View more versions of this product
Catalog No. S402891
Add to cart
Description
Description
Sturdy and nonabsorbent.
  • Supports sample
  • Smooth texture avoids trapping of material
  • Will not absorb moisture
  • Sold in packs of 500
  • Packaged in 4 mil poly bag for extra protection
Specifications
Specifications
Square
152 mm
Paper, Low Nitrogen
6 in.
6 in.
Weighing Paper
152 mm
6 x 6 in. (152 x 152mm)
For analytical balances