Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitromethane, 99%, Acros Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass Bottle
Metal drum
75-52-5
CH3NO2
61.04
LYGJENNIWJXYER-UHFFFAOYSA-N
methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4
6375
CHEBI:77701
nitromethane
C[N+](=O)[O-]
75-52-5
CH3NO2
61.04
LYGJENNIWJXYER-UHFFFAOYSA-N
methane, nitro,nitrocarbol,nitrometan,nitrometan polish,nitro-methane,unii-ru5wg8c3f4,ccris 1205,hsdb 106,meno2,ru5wg8c3f4
6375
CHEBI:77701
nitromethane
C[N+](=O)[O-]
Viscosity 0.61 mPa.s (25°C)
Packaging Glass Bottle
Infrared Spectrum Authentic
Melting Point -29.0°C
Color Undesignated
Flash Point 35°C
Refractive Index 1.3805 to 1.3825
Density 1.1200g/mL
Quantity 1L
Specific Gravity 1.12
Boiling Point 100.0°C to 102.0°C
Assay Percent Range 99%
Linear Formula CH3NO2
Beilstein 01,74
Fieser 01,739; 04,357; 09,323; 13,199
Merck Index 15,6698
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol,ether and dimethylformamide
Formula Weight 61.04
Physical Form Liquid
Percent Purity 99%
Grade Pure
Chemical Name or Material Nitromethane
Show More Show Less

;GHS P StatementKeep away from heat/sparks/open flames/hot surfaces.- No smoking.IF SWALLOWED: rinse mouth. Do NOT induce vomiting.Wash face,hands and any exposed skin thoroughly after handling.IF INHALED: Remove to fresh air and keep at