Alfa Aesar™ Nitromethane-d3, 99%(Isotopic)

Manufacturer: Alfa Aesar™ 4233814

Quantity 25g

 View more versions of this product

Catalog No. AA4233814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13031-32-8
    Formula Weight 64.06
    MDL Number MFCD00044214
    UN Number UN1261
    Chemical Name or Material Nitromethane-d3, Isotopic

    Specifications

    Formula Weight 64.06
    Refractive Index 1.3795
    UN Number UN1261
    Health Hazard 3 GHS P Statement
    P260-P370+P380+P375
    Do not breathe dust/fume/gas/mist/vapors/spray.
    In case of fire: Evacuate area.
    Fight fire remotely due to the risk of explosion.
    Boiling Point 100°C
    Physical Form Liquid
    MDL Number MFCD00044214
    CAS 13031-32-8
    Odor Characteristic
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: NITROMETHANE
    TSCA TSCA
    Density 1.183g/mL
    Quantity 25g
    Health Hazard 1 Warning
    Color Colorless
    Health Hazard 2 GHS H Statement
    H226-H302
    Flammable liquid and vapor.
    Harmful if swallowed.
    Chemical Name or Material Nitromethane-d3, Isotopic
    Assay 99%

    GHS H Statement
    H226-H302
    Flammable liquid and vapor.
    Harmful if swallowed.

    GHS P Statement
    P260-P370+P380+P375
    Do not breathe dust/fume/gas/mist/vapors/spray.
    In case of fire: Evacuate area.
    Fight fire remotely due to the risk of explosion.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit