Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitronium hexafluorophosphate, 95%, ACROS Organics™

Click to view available options
Quantity:
25g,25g
Packaging:
Plastic bottle,Plastic bottle
19200-21-6
F6NO2P
190.969
MFCD00031520
DXWZKTLGZWTYPL-UHFFFAOYSA-N
nitronium hexafluorophosphate,nitronium ion hexafluorophosphate,dioxidonitrogen 1+ hexafluorophosphate
45933661
nitronium;hexafluorophosphate
[N+](=O)=O.F[P-](F)(F)(F)(F)F
19200-21-6
F6NO2P
190.969
MFCD00031520
DXWZKTLGZWTYPL-UHFFFAOYSA-N
nitronium hexafluorophosphate,nitronium ion hexafluorophosphate,dioxidonitrogen 1+ hexafluorophosphate
45933661
nitronium;hexafluorophosphate
[N+](=O)=O.F[P-](F)(F)(F)(F)F
Quantity 25g,25g
Packaging Plastic bottle,Plastic bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 190.97
Chemical Name or Material Nitronium hexafluorophosphate

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.