Nitronium hexafluorophosphate, Alfa Aesar™

Manufacturer: Alfa Aesar 1315106

Quantity 5g

 View more versions of this product

Catalog No. AA1315106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 19200-21-6
    Physical Form Crystalline
    Molecular Formula NO2PF6
    Formula Weight 190.97
    MDL Number MFCD00031520
    Chemical Name or Material Nitronium hexafluorophosphate

    Specifications

    Physical Form Crystalline
    Formula Weight 190.97
    Quantity 5g
    Color Off-white
    DOT Information DOT Class: 8, Packaging Group: II
    EINECS Number 242-872-7
    Molecular Formula NO2PF6
    MDL Number MFCD00031520
    Appearance Off-white Cristals
    CAS 19200-21-6
    Health Hazard 1 H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.
    Chemical Name or Material Nitronium hexafluorophosphate

    H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit