Alfa Aesar™ Nitronium tetrafluoroborate, 0.3 to 0.5M soln. in sulfolane

Manufacturer: Alfa Aesar™ H34069AH

Quantity 200mL

 View more versions of this product

Catalog No. AAH34069AH

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13826-86-3
    Molecular Formula BF4NO2
    Linear Formula NO2BF4
    Formula Weight 132.81
    MDL Number MFCD00011432
    UN Number UN3265
    Chemical Name or Material Nitronium tetrafluoroborate

    Specifications

    Formula Weight 132.81
    TSCA TSCA
    Molecular Formula BF4NO2
    Linear Formula NO2BF4
    MDL Number MFCD00011432
    CAS 13826-86-3
    UN Number UN3265
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H334-H314
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.
    Health Hazard 3 GHS P Statement
    P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Concentration 0.3 to 0.5M soln. in sulfolane
    Quantity 200mL
    Chemical Name or Material Nitronium tetrafluoroborate

    GHS H Statement
    H334-H314
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Causes severe skin burns and eye damage.

    2° to 8°C

    GHS P Statement
    P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit