Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrosonium hexafluorophosphate, 95%, Acros Organics

Click to view available options
Quantity:
5g,5g
25g,25g
Packaging:
Plastic bottle,Plastic bottle
16921-91-8
F6NOP
174.97
MFCD00040324
SAKPNYRUBHJGAR-UHFFFAOYSA-N
nitrosonium hexafluorophosphate,nitrilooxonium hexafluorophosphate,nopf6,nitrosyl hexafluorophosphate,nitrosonium hexafluorophosphat,nitrosylhexafluorophosphate,azanylidyneoxidanium hexafluorophosphate,hexafluoro-$l^ 5-phosphanuide; nitrosonium
10910083
azanylidyneoxidanium;hexafluorophosphate
N#[O+].F[P-](F)(F)(F)(F)F
16921-91-8
F6NOP
174.97
MFCD00040324
SAKPNYRUBHJGAR-UHFFFAOYSA-N
nitrosonium hexafluorophosphate,nitrilooxonium hexafluorophosphate,nopf6,nitrosyl hexafluorophosphate,nitrosonium hexafluorophosphat,nitrosylhexafluorophosphate,azanylidyneoxidanium hexafluorophosphate,hexafluoro-$l^ 5-phosphanuide; nitrosonium
10910083
azanylidyneoxidanium;hexafluorophosphate
N#[O+].F[P-](F)(F)(F)(F)F
Color Colorless or White,Colorless or White
Quantity 25g,25g
Packaging Plastic bottle,Plastic bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min
Solubility Information Solubility in water: decomposes
Formula Weight 174.97
Physical Form Crystals
Percent Purity 95%
Chemical Name or Material Nitrosonium hexafluorophosphate
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 240-980-9