Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrosonium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Catalog Number AC174390050
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
14635-75-7
BF4HNO2
133.82
MFCD00011433
XYRQURYIJKJVSF-UHFFFAOYSA-N
nitrosonium tetrafluoroborate, nitrosyl tetrafluoroborate, nitrosyltetrafluoroborate, nobf4, nitrilooxonium tetrafluoroborate, azanylidyneoxidanium tetrafluoroborate
11137142
azanylidyneoxidanium;tetrafluoroborate
[O-][NH+]=O.F[B-](F)(F)F
14635-75-7
BF4HNO2
133.82
MFCD00011433
XYRQURYIJKJVSF-UHFFFAOYSA-N
nitrosonium tetrafluoroborate, nitrosyl tetrafluoroborate, nitrosyltetrafluoroborate, nobf4, nitrilooxonium tetrafluoroborate, azanylidyneoxidanium tetrafluoroborate
11137142
azanylidyneoxidanium;tetrafluoroborate
[O-][NH+]=O.F[B-](F)(F)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY
Melting Point 250.0°C
Color White
CAS Min % 96.0
CAS Max % 100.0
Physical Form Crystalline Powder and/or Crystals
Quantity 5 g
Assay Percent Range 96% min. (ex F)
Linear Formula NOBF4
Merck Index 15, 6736
Formula Weight 116.81
Percent Purity 97%
Packaging Plastic bottle
Specific Gravity 2.185
Density 2.1850g/mL
Chemical Name or Material Nitrosonium tetrafluoroborate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 238-679-2

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit