Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrosonium tetrafluoroborate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Plastic bottle
14635-75-7
BF4NO
116.81
MFCD00011433
KGCNVGDHOSFKFT-UHFFFAOYSA-N
nitrosonium tetrafluoroborate,nitrosyl tetrafluoroborate,nitrosyltetrafluoroborate,nobf4,nitrilooxonium tetrafluoroborate,azanylidyneoxidanium tetrafluoroborate
11137142
azanylidyneoxidanium;tetrafluoroborate
[B-](F)(F)(F)F.N#[O+]
14635-75-7
BF4NO
116.81
MFCD00011433
KGCNVGDHOSFKFT-UHFFFAOYSA-N
nitrosonium tetrafluoroborate,nitrosyl tetrafluoroborate,nitrosyltetrafluoroborate,nobf4,nitrilooxonium tetrafluoroborate,azanylidyneoxidanium tetrafluoroborate
11137142
azanylidyneoxidanium;tetrafluoroborate
[B-](F)(F)(F)F.N#[O+]
Oxidation reagent
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (ex F)
Linear Formula NOBF4
Merck Index 15, 6736
Formula Weight 116.81
Physical Form Crystals or Crystalline Powder
Percent Purity 97%
Density 2.1850g/mL
Packaging Plastic bottle
Specific Gravity 2.185
Color White
Melting Point 250.0°C
Quantity 25g
Chemical Name or Material Nitrosonium tetrafluoroborate
Show More Show Less

GHS H Statement
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 238-679-2