Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nitroterephthalic acid, 99%

Click to view available options
Quantity:
5g
25g
100g
610-29-7
C8H5NO6
211.129
MFCD00007141
QUMITRDILMWWBC-UHFFFAOYSA-N
nitroterephthalic acid,2-nitroterephthalic acid,2-nitrobenzene-1,4-dicarboxylic acid,1,4-benzenedicarboxylic acid, 2-nitro,terephthalic acid, nitro,2-nitroterephthalicacid,2-nitroterephthalate,5-nitroterephthalic acid,acmc-1au2u
69122
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O
610-29-7
C8H5NO6
211.129
MFCD00007141
QUMITRDILMWWBC-UHFFFAOYSA-N
nitroterephthalic acid,2-nitroterephthalic acid,2-nitrobenzene-1,4-dicarboxylic acid,1,4-benzenedicarboxylic acid, 2-nitro,terephthalic acid, nitro,2-nitroterephthalicacid,2-nitroterephthalate,5-nitroterephthalic acid,acmc-1au2u
69122
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O
Quantity 100g
Melting Point 270°C to 272°C
Formula Weight 211.13
Percent Purity 99%
Chemical Name or Material Nitroterephthalic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA