Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitroterephthalic acid, +99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
610-29-7
C8H5NO6
211.13
MFCD00007141
QUMITRDILMWWBC-UHFFFAOYSA-N
nitroterephthalic acid,2-nitroterephthalic acid,2-nitrobenzene-1,4-dicarboxylic acid,1,4-benzenedicarboxylic acid, 2-nitro,terephthalic acid, nitro,2-nitroterephthalicacid,2-nitroterephthalate,5-nitroterephthalic acid,acmc-1au2u
69122
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O
610-29-7
C8H5NO6
211.13
MFCD00007141
QUMITRDILMWWBC-UHFFFAOYSA-N
nitroterephthalic acid,2-nitroterephthalic acid,2-nitrobenzene-1,4-dicarboxylic acid,1,4-benzenedicarboxylic acid, 2-nitro,terephthalic acid, nitro,2-nitroterephthalicacid,2-nitroterephthalate,5-nitroterephthalic acid,acmc-1au2u
69122
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])C(=O)O
Infrared Spectrum Authentic
Color White to Beige
Packaging Glass bottle
Quantity 25g
Melting Point 270.0°C to 272.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Beilstein 09, 851
Formula Weight 211.13
Physical Form Powder
Percent Purity ≥99%
Chemical Name or Material Nitroterephthalic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning