Learn More
Nitrotryptophan Mouse anti-Chemical, Clone: 4F8, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527562
Catalog No. PIMA527562
Add to cart
Description
Description
6-nitrotryptophan (6-NO2-Trp) is shown to be one of the most stable and major ONOO- modified amino acids, and growing evidences suggest that nitrotryptophan may play a significant role in the cellular regulation under nitrosative stress. Reactive nitrogen species (RNS) are formed in a variety of diseases such as atherosclerosis, inflammation, cancer and neurodegenerative disiases. Peroxinitrite (ONOO-) is known as a potent biological oxidant, which is formed via very fast reaction between nitric oxide (NO) and superoxide (O2-). Peroxinitrite can induce nitration of aromatic amino acids such as Tyr, Trp, Phe and His, which results in modulation of protein functions.
Specifications
Specifications
Nitrotryptophan
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
6-Nitrotryptophan, Nitrotryptophan (NW), Nitrotryptophan, NW, 6-NO2-Trp
Synthetic 6-Nitrotryptophan modified Keyhole Limpet Hemocyanin (KLH)
Protein G
RUO
Antibody
Monoclonal
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
4F8
Unconjugated
Liquid
Mouse
IgG2b
100 μg
-20°C
Primary
Chemical