Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrotryptophan Mouse anti-Non-species specific, Clone: 2D12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259354

Catalog No. NBP259354


Add to cart

Description

Description

Nitrotryptophan Monoclonal antibody specifically detects Nitrotryptophan in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Nitrotryptophan
2D12
Unconjugated
PBS pH 7.4, 50% glycerol with 0.09% Sodium Azide
Mouse
IgG1
100 ug
Store at -20C.
Monoclonal
Non-species specific
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
1 mg/mL
Western Blot 1:1000, Flow Cytometry 1:50, ELISA 1:1000, Immunocytochemistry/Immunofluorescence 1:50
Purified
Synthetic 6-Nitrotryptophan modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
RUO
Primary
Specific for 6-Nitrotyrptophan modified proteins. Doss not cross-react with 7-Ketocholoesterol modified proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only