Learn More
Nitrotryptophan Mouse anti-Non-species specific, DyLight 680, Clone: 2D12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259354FR
Catalog No. NBP259354FR
Add to cart
Description
Description
Nitrotryptophan Monoclonal antibody specifically detects Nitrotryptophan in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Nitrotryptophan
2D12
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Purified
Synthetic 6-Nitrotryptophan modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG1
100 ul
Primary
Specific for 6-Nitrotyrptophan modified proteins. Doss not cross-react with 7-Ketocholoesterol modified proteins.