Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitrotryptophan Mouse anti-Nonspecies specific, PE/Cy5.5, Clone: 2D12, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259354PECY55

Catalog No. N259354PEC5


Add to cart

Description

Description

Nitrotryptophan Monoclonal antibody specifically detects Nitrotryptophan in Nonspecies specific samples. It is validated for Flow Cytometry.
Specifications

Specifications

Nitrotryptophan
2D12
Flow Cytometry
Conjugated, Purified
Synthetic 6-Nitrotryptophan modified Keyhole Limpet Hemocyanin (KLH).
Protein G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Nonspecies specific
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
Mouse
IgG1
0.1 mL
Primary
Specific for 6-Nitrotyrptophan modified proteins. Doss not cross-react with 7-Ketocholoesterol modified proteins.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit