Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nitrotyrosine Antibody (EM-30), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP196130AF488
Description
Nitrotyrosine Monoclonal specifically detects Nitrotyrosine in Human, Mouse, Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Nitrotyrosine | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
The antibody EM-30 detects nitrotyrosine group in biological material. It can be used as a marker for peroxynitrite formation in particular tissues | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
EM-30 | |
Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Nitrotyrosine | |
NO2-Tyr-CH2-Thyroglobulin | |
0.1 mL | |
Primary | |
Human, Non-species specific | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction