Learn More
Nitrotyrosine Mouse anti-Human, DyLight 680, Clone: EM-30, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196130FR
Catalog No. NBP196130FR
Add to cart
Description
Description
Nitrotyrosine Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
Nitrotyrosine
EM-30
Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Nitrotyrosine
NO2-Tyr-CH2-Thyroglobulin
Protein A or G purified
RUO
Primary
The antibody EM-30 detects nitrotyrosine group in biological material. It can be used as a marker for peroxynitrite formation in particular tissues