Learn More
Nitrotyrosine Mouse anti-Human, Nonspecies specific, Janelia Fluor 646, Clone: EM-30, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP196130JF646
Catalog No. NBP96130646
Add to cart
Description
Description
Nitrotyrosine Monoclonal antibody specifically detects Nitrotyrosine in Human, Nonspecies specific samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
Specifications
Nitrotyrosine
EM-30
Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
Mouse
IgG1
0.1mL
Primary
The antibody EM-30 detects nitrotyrosine group in biological material. It can be used as a marker for peroxynitrite formation in particular tissues
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Nitrotyrosine
NO2-Tyr-CH2-Thyroglobulin
Protein A purified
Store at 4°C in the dark.
Monoclonal
Human, Nonspecies specific