Learn More
Nitrotyrosine Mouse anti-Non-species specific, Alexa Fluor 647, Clone: 39B6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NB11096877AF647
Catalog No. NBT96877647
Add to cart
Description
Description
Nitrotyrosine Monoclonal antibody specifically detects Nitrotyrosine in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
Nitrotyrosine
39B6
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
Mouse
IgG2a
0.1 ml
Primary
Recognizes 3-nitrotyrosine moieties. No detectable cross-reactivity with non-nitrated tyrosine. Not species specific.
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Nitrotyrosine
Hybridoma line 39B6
Protein G purified
Store at 4C in the dark.
Monoclonal
Non-species specific