Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NK-4R Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537272
Description
This antibody detects endogenous protein at a molecular weight of 46 kDa. Purity is >95% by SDS-PAGE.
NK4R is a putative Tachykinin Receptor with sequence similarities to the neuromedin K receptor. It was initially thought to be an opioid receptor and is antagonized by the opioid peptide dynorphin. However, NK4R is activated by the tachykinin neurokinin B.
Specifications
NK-4R | |
Polyclonal | |
Unconjugated | |
Nk4 receptor; NK4R; Tac3rl; Tacr4 | |
A synthetic peptide corresponding to residues in human NK-4R. | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction