Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKAIN4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548959

Catalog No. PA548959

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

NKAIN4 is a member of a family of mammalian proteins (see NKAIN1; MIM 612871) with similarity to Drosophila Nkain and interacts with the beta subunit of Na,K-ATPase (ATP1B1; MIM 182330) (Gorokhova et al., 2007 [PubMed 17606467]).[supplied by OMIM, Jun 2009]
Specifications

Specifications

NKAIN4
Polyclonal
PBS with 0.09% sodium azide
Q8IVV8
Sodium/potassium-transporting ATPase subunit beta-1-interacting protein 4, Na(+)/K(+)-transporting ATPase subunit beta-1-interacting protein 4, Protein FAM77A, NKAIN4, C20orf58, FAM77A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
128414
Western Blot
Unconjugated
NKAIN4
Liquid
NKAIN4
KLH conjugated synthetic peptide between 167-195 amino acids from the C-terminal region of human NKAI4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit