Learn More
Invitrogen™ NKAIN4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113991
Description
Immunogen sequence: MGSCSGRCAL VVLCAFQLVA ALERQVFDFL GYQWAPILAN FVHIIIVILG For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 92%; Dog: 93%; Guinea Pig: 86%; Horse: 90%; Human: 100%; Mouse: 93%; Pig: 92%; Rabbit: 83%; Rat: 83%; Zebrafish: 86%.
NKAIN4 is a member of a family of mammalian proteins (see NKAIN1; MIM 612871) with similarity to Drosophila Nkain and interacts with the beta subunit of Na,K-ATPase (ATP1B1; MIM 182330) (Gorokhova et al., 2007 [PubMed 17606467]).[supplied by OMIM, Jun 2009].
Specifications
NKAIN4 | |
Polyclonal | |
Unconjugated | |
NKAIN4 | |
bA261N11.2; C20orf58; FAM77A; Na(+)/K(+)-transporting ATPase subunit beta-1-interacting protein 4; Na+/K+ transporting ATPase interacting 4; NKAIN4; Protein FAM77A; Sodium/potassium-transporting ATPase subunit beta-1-interacting protein 4 | |
Rabbit | |
Affinity chromatography | |
RUO | |
128414 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IVV8 | |
NKAIN4 | |
Synthetic peptide directed towards the N terminal region of human NKAIN4 (aa 1-50). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.