Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKCC1/SLC12A2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP159430
Description
NKCC1/SLC12A2 Polyclonal specifically detects NKCC1/SLC12A2 in Human samples. It is validated for Western Blot.Specifications
NKCC1/SLC12A2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
Basolateral Na-K-Cl symporter, BSC, BSC2, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1, NKCC1MGC104233, solute carrier family 12 (sodium/potassium/chloride transporters), member 2, solute carrier family 12 member 2 | |
Rabbit | |
131 kDa | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%; Guinea pig: 91%; Chicken: 78%. | |
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
P55011 | |
SLC12A2 | |
Synthetic peptides corresponding to SLC12A2(solute carrier family 12 (sodium/potassium/chloride transporters), member 2) The peptide sequence was selected from the C terminal of SLC12A2 (NP_001037). Peptide sequence IIAFEEIIEPYRLHEDDKEQDIADKMKEDEPWRITDNELELYKTKTYRQI The peptide sequence for this immunogen was taken from within the described region. | |
Protein A purified | |
RUO | |
6558 | |
Reconstitute with 100μL distilled water to a final antibody concentration of 1mg/mL. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction