Learn More
NKCC1/SLC12A2 Mouse anti-Human, Alexa Fluor 405, Clone: 915911, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8528V100UG
Catalog No. FAB8528V100
Add to cart
This item is not returnable. View return policy
Description
Description
NKCC1/SLC12A2 Monoclonal antibody specifically detects NKCC1/SLC12A2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
NKCC1/SLC12A2
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
P55011
SLC12A2
NS0 mouse myeloma cell line transfected with human NKCC1/SLC12A2 Met1-Ser1212 Accession # P55011
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains NKCC1/SLC12A2 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
915911
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Basolateral Na-K-Cl symporter, BSC, BSC2, Bumetanide-sensitive sodium-(potassium)-chloride cotransporter 1, NKCC1MGC104233, SLC12A2, solute carrier family 12 (sodium/potassium/chloride transporters), member 2, solute carrier family 12 member 2
Mouse
IgG2b
RUO
Primary
6558
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)