Learn More
NKG2A/C/E Rat anti-Mouse, BV605, Clone: 20d5, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 564382
Catalog No. BDB564382
Add to cart
Description
Description
NKG2A/C/E Monoclonal antibody specifically detects NKG2A/C/E in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
NKG2A/C/E
20d5
Brilliant Violet 605
Purified
Rat
Rat LEW
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV605 under optimum conditions
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
NKG2A/NKG2C/NKG2E; Klrc1/Klrc2/Klrc3; CD159a/CD159c/CD159e; CD159
Mouse CD94 and NKG2A Transfected Cell Line
50μg
Cell Biology
Primary
Murine
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.