Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NKG2D/CD314 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP243645
Description
Specifically detects NKG2D/CD314 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting
NKG2D/CD314 Polyclonal specifically detects NKG2D/CD314 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NKG2D/CD314 | |
Polyclonal | |
Unconjugated | |
PBS (pH7), 20% Glycerol with 0.025% Proclin 300 | |
KLRK1 | |
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human NKG2D. The exact sequence is proprietary. | |
Immunogen affinity purified | |
RUO | |
22914 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
Rabbit | |
25 kDa | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction