Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NKIAMRE Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120477
This item is not returnable.
View return policy
Description
Positive test controls include: HeLa, LO2, Mouse brain, Mouse liver, Rat brain. Immunogen sequence: MEMYETLGKV GEGSYGTVMK CKHKNTGQIV AIKIFYERPE QSVNKIAMRE IKFLKQFHHE NLVNLIEVFR QKKKIHLVFE FIDHTVLDEL QHYCHGLESK RLRKYLFQIL RAIDYLHSNN.
The protein encoded by this gene is a member of cyclin-dependent protein kinase (CDK) family. CDK family members are highly similar to the gene products of Saccharomyces cerevisiae cdc28, and Schizosaccharomyces pombe cdc2, and are known to be important regulators of cell cycle progression. This gene was identified as a gene absent in leukemic patients with chromosome 5q deletion. This loss may be an important determinant of dysmyelopoiesis. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
NKIAMRE | |
Polyclonal | |
Unconjugated | |
CDKL3 | |
AI852479; B230379H01Rik; CDKL3; cyclin dependent kinase like 3; cyclin-dependent kinase-like 3; NKIAMRE; Nkiatre; Nkiatreb; serine/threonine kinase NKIATRE beta; serine/threonine protein kinase NKIATRE; Serine/threonine-protein kinase NKIAMRE; serine-threonine protein kinase NKIAMRE | |
Rabbit | |
Affinity Chromatography | |
RUO | |
213084, 51265, 60396 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.37 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8BLF2, Q8IVW4, Q9JM01 | |
CDKL3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human CDKL3 (NP_001107047). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction