Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKIRAS1 Mouse anti-Human, Clone: 184C278, Abnova™

Mouse monoclonal antibody raised against synthetic peptide of human NKIRAS1.

Manufacturer:  Abnova Corporation MAB12954

Catalog No. 89-934-699


Add to cart

Description

Description

Sequence: RVELLKKEIDKFKDKKEV
Specifications

Specifications

NKIRAS1
184C278
Mouse monoclonal antibody raised against synthetic peptide of human NKIRAS1.
In PBS (0.02% sodium azide).
KBRAS1/kappaB-Ras1
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
NKIRAS1
A synthetic peptide corresponding to amino acids 96-113 of human NKIRAS1.
Protein G purification
RUO
Primary
28512
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit