Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NKp30 Mouse anti-Human, Clone: 30.95.1, MilliporeSigma™

Mouse Monoclonal Antibody

$153.40 - $387.00

Specifications

Antigen NKp30
Clone 30.95.1
Concentration 0.5 mg/mL
Applications Flow Cytometry
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF2241100
View Documents
MilliporeSigma
MABF2241100UL
100 μL
Each for $387.00
Add to cart
 
MABF224125
View Documents
MilliporeSigma
MABF224125UL
25 μL
Each for $153.40
Add to cart
 
Description

Description

NKp30 Monoclonal antibody specifically detects NKp30 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

NKp30
0.5 mg/mL
Unconjugated
Mouse
Human
Human NK cells.
Monoclonal
Protein G purified
30.95.1
Flow Cytometry
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
IgG1κ
Primary
Stable for 1 year at 2-8°C from date of receipt.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit